Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM129C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098524.1 | NP_001091994.1 | 5 | 620 | ||
NM_001321826.1 | NP_001308755.1 | 4 | 492 | |||
NM_001321827.1 | NP_001308756.1 | 4 | 492 | |||
NM_001321828.1 | NP_001308757.1 | 3 | 330 | |||
NM_173544.4 | NP_775815.2 | 5 | 620 | |||
XM_005259813.3 | XP_005259870.1 | 4 | 467 | |||
XM_005259814.3 | XP_005259871.1 | 4 | 467 | |||
XM_011527781.2 | XP_011526083.1 | 4 | 467 | |||
XM_011527786.2 | XP_011526088.1 | 4 | 467 | |||
XM_011527787.2 | XP_011526089.1 | 4 | 467 | |||
XM_011527789.1 | XP_011526091.1 | 4 | 467 | |||
XM_017026453.1 | XP_016881942.1 | 4 | 467 | |||
XM_017026454.1 | XP_016881943.1 | 4 | 467 | |||
XM_017026455.1 | XP_016881944.1 | 3 | 339 | |||
XM_017026456.1 | XP_016881945.1 | 3 | 394 | |||
GenBank mRNA | AK074069.1 | - | 4 | 442 | ||
AK097121.1 | - | 4 | 467 | |||
AK298062.1 | - | 3 | 321 | |||
AK300046.1 | - | 4 | 474 | |||
AK301159.1 | - | 4 | 370 | |||
AY254197.1 | - | 5 | 620 | |||
AY254198.1 | - | 5 | 620 | |||
AY254199.1 | - | 5 | 620 | |||
BC053606.1 | - | 4 | 426 | |||
DC296532.1 | - | 4 | 492 |