Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144757.1 | NP_001138229.1 | 6 | 752 | ||
NM_003020.3 | NP_003011.1 | 6 | 749 | |||
GenBank mRNA | BC005349.1 | - | 6 | 691 | 3949961 | |
BC093053.1 | - | 6 | 708 | 30720075 | ||
Y00757.1 | - | 6 | 660 |