Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003022.2 | NP_003013.1 | 4 | 1108 | ||
XM_011531013.1 | XP_011529315.1 | 5 | 912 | |||
XM_011531014.1 | XP_011529316.1 | 6 | 1231 | |||
GenBank mRNA | AF042081.1 | - | 4 | 789 | ||
AK024892.1 | - | 5 | 882 | |||
AK129610.1 | - | 4 | 793 | |||
AK310155.1 | - | 5 | 901 | |||
AK315897.1 | - | 4 | 773 | |||
AL136718.1 | - | 4 | 809 | |||
BC016709.1 | - | 4 | 814 | 4105289 | ||
BC103762.1 | - | 4 | 933 | 6179172 | ||
FJ224348.1 | - | 4 | 789 |