Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM126B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321618.1 | NP_001308547.1 | 11 | 3824 | ||
NM_001321619.1 | NP_001308548.1 | 12 | 4035 | |||
NM_001321621.1 | NP_001308550.1 | 11 | 3937 | |||
NM_001321622.1 | NP_001308551.1 | 13 | 4153 | |||
NM_001321623.1 | NP_001308552.1 | 13 | 4090 | |||
NM_001321624.1 | NP_001308553.1 | 12 | 3992 | |||
NM_001321625.1 | NP_001308554.1 | 13 | 4203 | |||
NM_001321626.1 | NP_001308555.1 | 13 | 4223 | |||
NM_001321627.1 | NP_001308556.1 | 14 | 4146 | |||
NM_001321628.1 | NP_001308557.1 | 14 | 4259 | |||
NM_001321629.1 | NP_001308558.1 | 13 | 3978 | |||
NM_173822.4 | NP_776183.1 | 12 | 3922 | |||
XM_017003879.1 | XP_016859368.1 | 14 | 4301 | |||
XM_017003881.1 | XP_016859370.1 | 12 | 3983 | |||
XM_017003882.1 | XP_016859371.1 | 14 | 4202 | |||
XM_017003883.1 | XP_016859372.1 | 13 | 4377 | |||
GenBank mRNA | AK001843.1 | - | 1 | 1354 | ||
AK027168.1 | - | 1 | 1794 | |||
AK097111.1 | - | 5 | 3166 | |||
AL832234.1 | - | 11 | 3781 | |||
AL832346.1 | - | 14 | 4146 | |||
AL833403.1 | - | 12 | 3906 | |||
BC039295.1 | - | 12 | 3918 | 5267063 |