Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317896.1 | NP_001304825.1 | 5 | 662 | ||
NM_001317897.1 | NP_001304826.1 | 4 | 371 | |||
NM_174921.2 | NP_777581.1 | 4 | 517 | |||
XM_011513640.2 | XP_011511942.1 | 4 | 650 | |||
XM_017007862.1 | XP_016863351.1 | 5 | 566 | |||
XM_017007863.1 | XP_016863352.1 | 4 | 407 | |||
GenBank mRNA | AK023351.1 | - | 4 | 380 | ||
BC008502.1 | - | 4 | 380 | 4295121 | ||
BQ049060.1 | - | 4 | 362 | |||
HY155982.1 | - | 4 | 518 |