Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057592.1 | - | 1 | 1258 | ||
AK057596.1 | - | 1 | 1935 | |||
AK093195.1 | - | 1 | 751 | |||
BC117383.1 | - | 1 | 1901 | |||
BC126153.1 | - | 1 | 1898 |
Gene Symbol: LOC100506076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103733.1 | 1 | 68 |
Gene Symbol: LOC100506123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130704.1 | 1 | 68 |