Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003032.2 | NP_003023.1 | 7 | 3555 | ||
NM_173216.2 | NP_775323.1 | 8 | 3878 | |||
NM_173217.2 | NP_775324.1 | 7 | 3221 | |||
XM_005247719.1 | XP_005247776.1 | 7 | 3466 | |||
XM_005247720.1 | XP_005247777.1 | 7 | 3524 | |||
XM_006713734.1 | XP_006713797.1 | 9 | 4133 | |||
XM_011513085.1 | XP_011511387.1 | 9 | 4141 | |||
XM_011513086.1 | XP_011511388.1 | 7 | 3524 | |||
XM_017007065.1 | XP_016862554.1 | 10 | 4225 | |||
XM_017007066.1 | XP_016862555.1 | 7 | 3553 | |||
XM_017007067.1 | XP_016862556.1 | 7 | 3673 | |||
XM_017007068.1 | XP_016862557.1 | 7 | 3558 | |||
GenBank mRNA | AF007133.1 | - | 1 | 1399 | ||
AK128726.1 | - | 3 | 4169 | |||
BC031476.1 | - | 7 | 3549 | 4339124 | ||
BC040009.1 | - | 7 | 3555 | 5557868 |