Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC031256.1 | - | 7 | 1153 | 5271599 | |
BC047510.1 | - | 6 | 1007 | 5297045 |