Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199511.2 | NP_955805.1 | 2 | 2510 | ||
NM_199512.2 | NP_955806.1 | 2 | 1913 | |||
GenBank mRNA | AB052098.1 | - | 2 | 2127 | ||
AB266387.1 | - | 2 | 2698 | |||
AF506819.1 | - | 2 | 1827 | |||
AK075210.1 | - | 1 | 1748 | |||
AK292881.1 | - | 2 | 2194 | |||
AK304561.1 | - | 3 | 797 | |||
AL833034.1 | - | 2 | 1781 | |||
AY333429.1 | - | 2 | 2407 | |||
AY526612.1 | - | 2 | 2498 | |||
AY548106.1 | - | 2 | 2483 | |||
AY548107.1 | - | 2 | 2483 | |||
AY726593.1 | - | 2 | 1964 | |||
BC110842.1 | - | 2 | 2495 | 6205672 | ||
BC116178.1 | - | 2 | 2400 | 40068210 | ||
BX647117.1 | - | 2 | 2384 | |||
BX647817.1 | - | 2 | 1799 |