Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178554.4 | NP_848649.3 | 11 | 2008 | ||
XM_005247418.3 | XP_005247475.1 | 10 | 6976 | |||
XM_006713612.3 | XP_006713675.1 | 10 | 6922 | |||
XM_017006287.1 | XP_016861776.1 | 11 | 7054 | |||
XM_017006288.1 | XP_016861777.1 | 11 | 7000 | |||
XM_017006289.1 | XP_016861778.1 | 10 | 6991 | |||
XM_017006290.1 | XP_016861779.1 | 10 | 6937 | |||
GenBank mRNA | AF086360.1 | - | 1 | 272 | ||
AK090526.1 | - | 12 | 1708 | |||
AK293840.1 | - | 10 | 1941 | |||
BC136349.1 | - | 10 | 1778 |