Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256440.1 | NP_001243369.1 | 9 | 1256 | ||
NM_001256441.1 | NP_001243370.1 | 9 | 1251 | |||
NR_046202.1 | 9 | 1304 | ||||
XM_005258520.2 | XP_005258577.1 | 9 | 1245 | |||
XM_017026291.1 | XP_016881780.1 | 9 | 1256 | |||
XM_017026294.1 | XP_016881783.1 | 6 | 974 | |||
XM_017026295.1 | XP_016881784.1 | 10 | 1354 | |||
GenBank mRNA | AK094207.1 | - | 9 | 1244 | ||
AL834131.1 | - | 9 | 1281 | |||
BC023623.2 | - | 9 | 1282 | 4662735 | ||
BC027935.1 | - | 9 | 1272 | 5202287 | ||
CR936760.1 | - | 9 | 1257 |