Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178839.4 | NP_849161.2 | 2 | 2068 | ||
GenBank mRNA | AK056644.1 | - | 2 | 2099 | ||
AK225779.1 | - | 2 | 2455 | |||
AK290512.1 | - | 2 | 2318 | |||
AY358310.1 | - | 2 | 2517 | |||
BC045113.1 | - | 2 | 2068 | 5200499 |