Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STT3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178862.2 | NP_849193.1 | 1, 2 | 815 | ||
XM_011533465.1 | XP_011531767.1 | 1, 2 | 678 | |||
XM_017005857.1 | XP_016861346.1 | 1, 2 | 678 | |||
GenBank mRNA | AK074587.1 | - | 1, 2 | 344 | ||
AK311488.1 | - | 1, 2 | 678 | |||
AX799082.1 | - | 1, 2 | 317 | |||
AY074880.1 | - | 1, 2 | 317 |