Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182501.3 | NP_872307.2 | 4 | 1239 | ||
NR_028049.1 | 4 | 1239 | ||||
XM_011510634.2 | XP_011508936.1 | 4 | 1496 | |||
XM_017003379.1 | XP_016858868.1 | 5 | 1473 | |||
XM_017003380.1 | XP_016858869.1 | 5 | 1473 | |||
XM_017003381.1 | XP_016858870.1 | 5 | 1473 | |||
XM_017003382.1 | XP_016858871.1 | 5 | 1473 | |||
XM_017003383.1 | XP_016858872.1 | 5 | 1473 | |||
XM_017003384.1 | XP_016858873.1 | 5 | 1473 | |||
XM_017003385.1 | XP_016858874.1 | 5 | 1473 | |||
XM_017003386.1 | XP_016858875.1 | 5 | 1944 | |||
XM_017003387.1 | XP_016858876.1 | 4 | 1496 | |||
XM_017003388.1 | XP_016858877.1 | 4 | 1496 | |||
XM_017003390.1 | XP_016858879.1 | 3 | 729 | |||
XM_017003391.1 | XP_016858880.1 | 3 | 1749 | |||
GenBank mRNA | AK055474.1 | - | 1 | 107 | ||
AK131298.1 | - | 4 | 2013 | |||
AK291665.1 | - | 4 | 1194 | |||
BC053533.1 | - | 4 | 1192 | 4417008 | ||
BX648184.1 | - | 4 | 1205 |