Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199692.1 | NP_001186621.1 | 7, 8 | 2014 | ||
NM_001199693.1 | NP_001186622.1 | 6, 7 | 1040 | |||
NM_001199694.1 | NP_001186623.1 | 6, 7 | 1461 | |||
NM_003040.3 | NP_003031.3 | 7, 8 | 1230 | |||
XM_006716094.3 | XP_006716157.1 | 7, 8 | 1230 | |||
GenBank mRNA | AB209158.1 | - | 7, 8 | 2014 | ||
AK295767.1 | - | 6, 7 | 1040 | |||
AK312699.1 | - | 7, 8 | 1209 | |||
BC004893.2 | - | 7, 8 | 867 | 3507709 | ||
BC009386.2 | - | 6, 7 | 1461 | 4127724 | ||
BC009434.1 | - | 6, 7 | 1461 | 3501642 | ||
BC010069.2 | - | 7, 8 | 867 | 4125444 | ||
BC028601.2 | - | 6, 7 | 1062 | 4814066 | ||
U62531.1 | - | 7, 8 | 1148 | |||
X62137.1 | - | 7, 8 | 1053 |