Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304366.1 | NP_001291295.1 | 9 | 2216 | ||
NM_182610.3 | NP_872416.1 | 9 | 2239 | |||
NR_130713.1 | 10 | 2329 | ||||
GenBank mRNA | BX537903.1 | - | 9 | 2119 | ||
BX648534.1 | - | 9 | 2097 | |||
BX648989.1 | - | 9 | 2119 | |||
BX648991.1 | - | 9 | 2216 |