Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182627.1 | NP_872433.1 | 4 | 984 | ||
GenBank mRNA | AK314105.1 | - | 4 | 993 | ||
BC054030.1 | - | 4 | 984 | 6145598 | ||
EF011620.1 | - | 4 | 984 |