Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182628.2 | NP_872434.2 | 8 | 814 | ||
XM_006713623.2 | XP_006713686.1 | 9 | 965 | |||
XM_017006320.1 | XP_016861809.1 | 9 | 965 | |||
XM_017006321.1 | XP_016861810.1 | 9 | 965 | |||
XM_017006322.1 | XP_016861811.1 | 9 | 965 | |||
XM_017006323.1 | XP_016861812.1 | 9 | 965 | |||
XM_017006324.1 | XP_016861813.1 | 9 | 965 | |||
XM_017006325.1 | XP_016861814.1 | 9 | 965 | |||
XM_017006326.1 | XP_016861815.1 | 9 | 965 | |||
XM_017006327.1 | XP_016861816.1 | 9 | 965 | |||
GenBank mRNA | AK097402.1 | - | 8 | 814 | ||
BC101366.1 | - | 7 | 747 | 40025823 | ||
BC101367.2 | - | 7 | 744 | 40025824 | ||
BC101368.1 | - | 7 | 744 | 40025828 | ||
BC101369.1 | - | 7 | 744 | 40025829 |