Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008779.1 | NP_001008779.1 | 4 | 445 | ||
NM_001142634.1 | NP_001136106.1 | 5 | 559 | |||
NM_182756.3 | NP_877433.2 | 6 | 633 | |||
GenBank mRNA | AY820303.1 | - | 4 | 427 | ||
AY820304.1 | - | 4 | 427 | |||
BC093005.1 | - | 5 | 559 | 4839330 |