Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166496.1 | NP_001159968.1 | 5 | 3555 | ||
NM_003051.3 | NP_003042.3 | 5 | 3086 | |||
XM_011542026.2 | XP_011540328.1 | 5 | 2819 | |||
XM_011542027.2 | XP_011540329.1 | 5 | 2829 | |||
GenBank mRNA | AJ438942.2 | - | 1 | 1425 | ||
AK000641.1 | - | 2 | 1971 | |||
AL162079.1 | - | 5 | 2916 | |||
BC026317.1 | - | 5 | 2929 | 4811581 |