Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194247.2 | NP_919223.1 | 6 | 766 | ||
XM_005246380.1 | XP_005246437.1 | 6 | 766 | |||
XM_005246381.4 | XP_005246438.1 | 6 | 672 | |||
XM_006712365.2 | XP_006712428.1 | 6 | 766 | |||
XM_017003587.1 | XP_016859076.1 | 6 | 672 | |||
XM_017003588.1 | XP_016859077.1 | 6 | 766 | |||
GenBank mRNA | AK293125.1 | - | 6 | 634 | ||
AK304687.1 | - | 6 | 629 | |||
AK316434.1 | - | 6 | 634 | |||
AY363225.1 | - | 6 | 660 | |||
BC064494.1 | - | 6 | 678 | |||
BC080591.1 | - | 2 | 94 | |||
BC112027.1 | - | 6 | 688 | 8327495 | ||
BC113470.1 | - | 6 | 688 | 8322522 | ||
BX647217.1 | - | 6 | 623 |