Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115007.2 | NP_001108479.1 | 12 | 1334 | ||
NM_001115008.2 | NP_001108480.1 | 12 | 1535 | |||
NM_001288996.1 | NP_001275925.1 | 9 | 1931 | |||
NM_001288997.1 | NP_001275926.1 | 12 | 1272 | |||
NM_194282.3 | NP_919258.2 | 12 | 2198 | |||
NR_110253.1 | 9 | 1857 | ||||
NR_110254.1 | 11 | 1531 | ||||
XM_005262750.4 | XP_005262807.1 | 11 | 2007 | |||
XM_006714081.3 | XP_006714144.1 | 11 | 1933 | |||
XM_017007728.1 | XP_016863217.1 | 9 | 1740 | |||
XM_017007729.1 | XP_016863218.1 | 10 | 1923 | |||
XM_017007730.1 | XP_016863219.1 | 10 | 1312 | |||
GenBank mRNA | AB111889.1 | - | 11 | 1518 | ||
AK292769.1 | - | 12 | 1282 | |||
AK294256.1 | - | 12 | 2127 | |||
AK299787.1 | - | 7 | 740 | |||
BC109277.2 | - | 9 | 1733 | |||
BC109278.2 | - | 9 | 1806 | 40028371 | ||
BX537919.1 | - | 11 | 1432 | |||
BX640657.1 | - | 12 | 1261 | |||
BX640966.1 | - | 12 | 2193 |