Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195833.1 | NP_001182762.1 | 10 | 1538 | ||
NM_198445.3 | NP_940847.1 | 10 | 1522 | |||
XM_006723015.2 | XP_006723078.1 | 10 | 1532 | |||
XM_006723016.2 | XP_006723079.1 | 10 | 1500 | |||
XM_011526454.2 | XP_011524756.1 | 10 | 1602 | |||
XM_011526455.2 | XP_011524757.1 | 11 | 1436 | |||
GenBank mRNA | AK124506.1 | - | 2 | 146 | ||
AK126119.1 | - | 7 | 1918 | |||
AK127808.1 | - | 10 | 1497 | |||
AK298327.1 | - | 10 | 1538 | |||
BU196609.1 | - | 1 | 92 |