Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198504.2 | NP_940906.1 | 1 | 1265 | ||
GenBank mRNA | AK123932.1 | - | 1 | 1265 |
Set Membership: |
G-protein coupled receptor |