Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005669.4 | NP_005660.4 | 2 | 332 | ||
XM_017009844.1 | XP_016865333.1 | 2 | 249 | |||
GenBank mRNA | AK294496.1 | - | 1 | 318 | ||
AK295436.1 | - | 2 | 223 | |||
AK298268.1 | - | 2 | 236 | |||
AK299902.1 | - | 2 | 228 | |||
AK310729.1 | - | 1 | 328 | |||
AK312522.1 | - | 2 | 237 | |||
AY562243.1 | - | 2 | 188 | |||
BC000232.1 | - | 2 | 213 | 3350749 | ||
BC065926.1 | - | 2 | 191 | 6164621 | ||
BX648005.1 | - | 2 | 313 | |||
DA807043.1 | - | 2 | 333 | |||
M74090.1 | - | 2 | 214 |