Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005669.4 | NP_005660.4 | 3 | 372 | ||
XM_017009844.1 | XP_016865333.1 | 3 | 289 | |||
GenBank mRNA | AK294496.1 | - | 2 | 358 | ||
AK298268.1 | - | 3 | 276 | |||
AK299902.1 | - | 3 | 268 | |||
AK310729.1 | - | 2 | 368 | |||
AK312522.1 | - | 3 | 277 | |||
AY562243.1 | - | 3 | 228 | |||
BC000232.1 | - | 3 | 253 | 3350749 | ||
BC065926.1 | - | 3 | 231 | 6164621 | ||
BX648005.1 | - | 3 | 353 | |||
DA807043.1 | - | 3 | 373 | |||
M74090.1 | - | 3 | 254 |