Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005754.2 | NP_005745.1 | 7 | 815 | ||
NM_198395.1 | NP_938405.1 | 7 | 784 | |||
XM_006714749.3 | XP_006714812.1 | 7 | 804 | |||
XM_006714750.3 | XP_006714813.1 | 7 | 743 | |||
XM_017008922.1 | XP_016864411.1 | 7 | 1090 | |||
XM_017008923.1 | XP_016864412.1 | 7 | 1059 | |||
GenBank mRNA | AK130003.1 | - | 329 | |||
AK222606.1 | - | 7 | 776 | |||
AK300098.1 | - | 6 | 685 | |||
AK303580.1 | - | 7 | 872 | |||
AK312572.1 | - | 7 | 778 | |||
BC006997.1 | - | 7 | 778 | 3950738 | ||
BC108278.1 | - | 7 | 723 | 6051310 | ||
BT019384.1 | - | 6 | 644 | |||
BX647869.1 | - | 6 | 686 | |||
BX648012.1 | - | 7 | 790 | |||
CR536535.1 | - | 6 | 644 | |||
U32519.1 | - | 7 | 765 |