Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270660.1 | NP_001257589.1 | 11 | 1131 | ||
NM_001270661.1 | NP_001257590.1 | 9 | 960 | |||
NM_005885.3 | NP_005876.2 | 12 | 1275 | |||
XM_011513932.2 | XP_011512234.1 | 12 | 1033 | |||
XM_011513934.1 | XP_011512236.1 | 12 | 1236 | |||
XM_011513936.2 | XP_011512238.1 | 12 | 1236 | |||
XM_011513937.2 | XP_011512239.1 | 2 | 617 | |||
XM_017008944.1 | XP_016864433.1 | 12 | 1236 | |||
GenBank mRNA | AB011169.1 | - | 12 | 1198 | ||
AK296585.1 | - | 9 | 883 | |||
AK300034.1 | - | 11 | 1067 | |||
BC046148.1 | - | 4 | 192 | |||
BC136461.1 | - | 12 | 1147 | 9020450 | ||
BC136462.1 | - | 12 | 1147 | 9020451 | ||
BC142679.1 | - | 12 | 1126 | 40148250 | ||
BC142694.1 | - | 12 | 1126 | 40148713 |