Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLU7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006425.4 | NP_006416.3 | 13, 14 | 1546 | ||
XM_005265793.3 | XP_005265850.1 | 13, 14 | 1893 | |||
XM_011534418.2 | XP_011532720.1 | 14, 15 | 1596 | |||
GenBank mRNA | AF101074.1 | - | 12, 13 | 1391 | ||
AK000813.1 | - | 3, 4 | 298 | |||
AK024875.1 | - | 13, 14 | 1466 | |||
AK314564.1 | - | 13, 14 | 1466 | |||
BC010634.1 | - | 13, 14 | 1472 | 3871882 |