Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006547.2 | NP_006538.2 | 5, 6 | 668 | ||
XM_006715639.2 | XP_006715702.1 | 4, 5 | 296 | |||
XM_011515089.2 | XP_011513391.1 | 6, 7 | 981 | |||
XM_011515092.2 | XP_011513394.1 | 6, 7 | 354 | |||
XM_011515093.2 | XP_011513395.1 | 5, 6 | 226 | |||
GenBank mRNA | AF117108.1 | - | 5, 6 | 668 | ||
BC065269.1 | - | 5, 6 | 647 | 6154021 | ||
U76705.1 | - | 5, 6 | 652 | |||
U97188.1 | - | 5, 6 | 652 |