Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006547.2 | NP_006538.2 | 15 | 2557 | ||
XM_006715639.2 | XP_006715702.1 | 14 | 2185 | |||
XM_011515089.2 | XP_011513391.1 | 16 | 2870 | |||
XM_011515090.2 | XP_011513392.1 | 12 | 3436 | |||
XM_011515091.2 | XP_011513393.1 | 13 | 2502 | |||
XM_011515092.2 | XP_011513394.1 | 16 | 2243 | |||
XM_011515093.2 | XP_011513395.1 | 15 | 2115 | |||
GenBank mRNA | AF117108.1 | - | 15 | 2557 | ||
BX640800.1 | - | 9 | 1568 | |||
BX648488.1 | - | 8 | 1440 | |||
U76705.1 | - | 15 | 2541 | |||
U97188.1 | - | 15 | 2541 |