Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007013.3 | NP_008944.1 | 5 | 539 | ||
XM_005250760.3 | XP_005250817.1 | 4 | 500 | |||
XM_005250761.3 | XP_005250818.1 | 5 | 583 | |||
XM_011516802.2 | XP_011515104.1 | 4 | 300 | |||
XM_017012993.1 | XP_016868482.1 | 4 | 495 | |||
XM_017012994.1 | XP_016868483.1 | 4 | 490 | |||
XM_017012995.1 | XP_016868484.1 | 5 | 566 | |||
XM_017012996.1 | XP_016868485.1 | 4 | 484 | |||
XM_017012997.1 | XP_016868486.1 | 4 | 463 | |||
GenBank mRNA | AK292800.1 | - | 5 | 554 | ||
AK310683.1 | - | 2 | 98 | |||
AL136739.1 | - | 3 | 269 | |||
AY043361.1 | - | 4 | 344 | |||
AY345857.1 | - | 5 | 573 | |||
BC036065.1 | - | 5 | 543 | 5296005 |