Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007468.2 | NP_001007469.1 | 9 | 1661 | ||
NM_001317946.1 | NP_001304875.1 | 9 | 1715 | |||
NM_003073.4 | NP_003064.2 | 9 | 1688 | |||
XM_011530345.2 | XP_011528647.1 | 9 | 1711 | |||
GenBank mRNA | AJ011737.1 | - | 9 | 1597 | ||
AJ011738.1 | - | 9 | 1569 | |||
AK021419.1 | - | 9 | 1594 | |||
AK024025.1 | - | 9 | 1683 | |||
AL389878.1 | - | 6 | 651 | |||
AL389881.1 | - | 2 | 469 |