Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012382.2 | NP_036514.1 | 4 | 559 | ||
XM_011514003.2 | XP_011512305.1 | 4 | 1684 | |||
GenBank mRNA | AF023244.1 | - | 4 | 505 | ||
AK303119.1 | - | 3 | 467 | |||
AK312560.1 | - | 4 | 533 | |||
BC015701.1 | - | 4 | 529 | 3907836 | ||
BC036536.1 | - | 4 | 508 | 5262401 | ||
DA518274.1 | - | 4 | 559 |