Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130420.1 | NP_001123892.1 | 9 | 876 | ||
NM_003075.3 | NP_003066.2 | 9 | 876 | |||
NM_139067.2 | NP_620706.1 | 9 | 876 | |||
XM_005269101.1 | XP_005269158.1 | 9 | 813 | |||
XM_005269102.1 | XP_005269159.1 | 9 | 813 | |||
XM_005269103.1 | XP_005269160.1 | 9 | 813 | |||
XM_005269104.1 | XP_005269161.1 | 9 | 813 | |||
XM_011538693.2 | XP_011536995.1 | 8 | 722 | |||
XM_017019884.1 | XP_016875373.1 | 9 | 813 | |||
XM_017019885.1 | XP_016875374.1 | 9 | 813 | |||
XM_017019886.1 | XP_016875375.1 | 9 | 813 | |||
XM_017019887.1 | XP_016875376.1 | 8 | 700 | |||
GenBank mRNA | AB209006.1 | - | 9 | 876 | ||
AB209293.1 | - | 9 | 785 | |||
AK293896.1 | - | 9 | 763 | |||
AK315153.1 | - | 9 | 776 | |||
BC013045.1 | - | 9 | 802 | 4180388 | ||
BC026222.2 | - | 9 | 795 | 4395012 | ||
BT009924.1 | - | 9 | 770 | |||
U66616.1 | - | 9 | 792 |