Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014149.3 | NP_054868.3 | 5 | 730 | ||
XM_011516083.2 | XP_011514385.1 | 5 | 700 | |||
XM_011516086.2 | XP_011514388.1 | 5 | 700 | |||
XM_017012070.1 | XP_016867559.1 | 5 | 700 | |||
XM_017012071.1 | XP_016867560.1 | 2 | 103 | |||
XM_017012072.1 | XP_016867561.1 | 5 | 680 | |||
GenBank mRNA | AF161534.1 | - | 5 | 672 | ||
AK001606.1 | - | 5 | 685 | |||
AK022935.1 | - | 5 | 674 | |||
AL136940.1 | - | 5 | 581 | |||
BC009939.2 | - | 3 | 188 | |||
BC017246.2 | - | 5 | 669 | 4890904 | ||
BC063394.1 | - | 5 | 666 | |||
BX640957.1 | - | 5 | 701 | |||
CR533561.1 | - | 4 | 512 |