Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014149.3 | NP_054868.3 | 15 | 3051 | ||
XM_011516083.2 | XP_011514385.1 | 16 | 3156 | |||
XM_011516086.2 | XP_011514388.1 | 16 | 3078 | |||
XM_017012070.1 | XP_016867559.1 | 15 | 3099 | |||
XM_017012071.1 | XP_016867560.1 | 13 | 2591 | |||
XM_017012072.1 | XP_016867561.1 | 15 | 3033 | |||
GenBank mRNA | AK001606.1 | - | 15 | 3006 | ||
AK074466.1 | - | 8 | 1811 | |||
AK124765.1 | - | 1 | 3786 | |||
AL136940.1 | - | 15 | 2902 | |||
BC009939.2 | - | 13 | 2509 |