Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145838.1 | NP_001139310.1 | 3, 4 | 506 | ||
NM_001145839.1 | NP_001139311.1 | 5, 6 | 737 | |||
NM_014637.3 | NP_055452.3 | 5, 6 | 737 | |||
XM_006716484.2 | XP_006716547.2 | 5, 6 | 709 | |||
XM_011517626.1 | XP_011515928.1 | 5, 6 | 691 | |||
XM_011517627.2 | XP_011515929.1 | 5, 6 | 627 | |||
XM_011517628.2 | XP_011515930.1 | 4, 5 | 844 | |||
GenBank mRNA | AK304716.1 | - | 5, 6 | 651 | ||
BC036116.1 | - | 5, 6 | 651 | 5299040 | ||
BC043498.2 | - | 5, 6 | 636 | 6057524 | ||
BX537854.1 | - | 3, 4 | 398 | |||
BX647507.1 | - | 5, 6 | 656 | |||
CR456910.1 | - | 3, 4 | 468 | |||
D13634.1 | - | 4, 5 | 485 |