Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014886.4 | NP_055701.1 | 5 | 919 | ||
NR_073403.1 | 6 | 1028 | ||||
XM_011543098.1 | XP_011541400.1 | 5 | 597 | |||
GenBank mRNA | AF077615.1 | - | 5 | 633 | ||
AF372458.1 | - | 5 | 644 | |||
AJ012409.1 | - | 5 | 631 | |||
AK313784.1 | - | 5 | 662 | |||
AY443419.1 | - | 4 | 549 | |||
BC005288.1 | - | 5 | 662 | 3683720 |