Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015089.3 | NP_055904.1 | 11 | 2682 | ||
XM_011514422.1 | XP_011512724.1 | 11 | 2691 | |||
XM_011514423.2 | XP_011512725.1 | 12 | 2793 | |||
XM_011514424.1 | XP_011512726.1 | 11 | 2691 | |||
XM_011514425.1 | XP_011512727.1 | 9 | 2361 | |||
XM_011514426.1 | XP_011512728.1 | 11 | 2691 | |||
XM_011514427.1 | XP_011512729.1 | 7 | 1310 | |||
XM_011514428.1 | XP_011512730.1 | 11 | 2691 | |||
XM_011514429.1 | XP_011512731.1 | 4 | 515 | |||
XM_011514430.1 | XP_011512732.1 | 11 | 2691 | |||
XM_017010589.1 | XP_016866078.1 | 11 | 3402 | |||
XM_017010590.1 | XP_016866079.1 | 5 | 864 | |||
GenBank mRNA | AB014608.3 | - | 3 | 316 | ||
AJ318215.1 | - | 10 | 2607 | |||
AY145132.1 | - | 10 | 2607 | |||
CR749511.1 | - | 10 | 3061 | |||
CR749841.1 | - | 11 | 2681 |