Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015089.3 | NP_055904.1 | 36 | 6956 | ||
XM_011514422.1 | XP_011512724.1 | 35 | 7112 | |||
XM_011514423.2 | XP_011512725.1 | 36 | 7214 | |||
XM_011514424.1 | XP_011512726.1 | 35 | 7031 | |||
XM_011514425.1 | XP_011512727.1 | 33 | 6782 | |||
XM_011514426.1 | XP_011512728.1 | 35 | 7135 | |||
XM_011514427.1 | XP_011512729.1 | 31 | 5731 | |||
XM_011514429.1 | XP_011512731.1 | 28 | 4936 | |||
XM_017010589.1 | XP_016866078.1 | 35 | 7823 | |||
XM_017010590.1 | XP_016866079.1 | 29 | 5285 | |||
GenBank mRNA | AB014608.3 | - | 28 | 4590 | ||
AJ318215.1 | - | 35 | 6881 | |||
AK128364.1 | - | 1 | 846 | |||
AY145132.1 | - | 35 | 6881 | |||
BC002879.2 | - | 13 | 2222 | |||
CR749511.1 | - | 35 | 7335 | |||
CR749841.1 | - | 36 | 6871 |