Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIP5K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276277.1 | NP_001263206.1 | 3 | 652 | ||
NM_001281471.1 | NP_001268400.1 | 3 | 652 | |||
NM_015216.3 | NP_056031.2 | 2 | 417 | |||
XM_005271935.1 | XP_005271992.1 | 3 | 651 | |||
XM_005271936.1 | XP_005271993.1 | 3 | 651 | |||
XM_005271938.1 | XP_005271995.1 | 3 | 651 | |||
XM_005277534.1 | XP_005277591.1 | 3 | 651 | |||
XM_005277535.1 | XP_005277592.1 | 3 | 651 | |||
XM_005277536.1 | XP_005277593.1 | 3 | 651 | |||
XM_005277538.1 | XP_005277595.1 | 3 | 651 | |||
XM_005277539.1 | XP_005277596.1 | 3 | 651 | |||
XM_005277541.1 | XP_005277598.1 | 3 | 651 | |||
XM_006714576.1 | XP_006714639.1 | 3 | 651 | |||
XM_006714577.1 | XP_006714640.1 | 3 | 651 | |||
XM_011543290.2 | XP_011541592.1 | 3 | 651 | |||
XM_017009269.1 | XP_016864758.1 | 3 | 651 | |||
XM_017009270.1 | XP_016864759.1 | 3 | 651 | |||
XM_017009271.1 | XP_016864760.1 | 3 | 651 | |||
XM_017009272.1 | XP_016864761.1 | 3 | 651 | |||
XM_017009273.1 | XP_016864762.1 | 3 | 651 | |||
XM_017009274.1 | XP_016864763.1 | 3 | 651 | |||
XM_017009275.1 | XP_016864764.1 | 3 | 651 | |||
GenBank mRNA | AB007893.1 | - | 3 | 642 | ||
AK294787.1 | - | 3 | 516 | |||
BC024591.1 | - | 2 | 170 | 3913521 |
Gene Symbol: SLC44A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936677.1 | - | 4 | 758 |