Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015310.3 | NP_056125.3 | 14 | 2770 | ||
NM_206909.2 | NP_996792.1 | 11 | 1174 | |||
XM_017013258.1 | XP_016868747.1 | 14 | 4079 | |||
XM_017013259.1 | XP_016868748.1 | 14 | 4057 | |||
XM_017013260.1 | XP_016868749.1 | 14 | 2830 | |||
XM_017013261.1 | XP_016868750.1 | 13 | 3044 | |||
XM_017013265.1 | XP_016868754.1 | 11 | 1580 | |||
GenBank mRNA | AB023159.2 | - | 11 | 1155 | ||
AF243495.2 | - | 12 | 1180 | |||
AF519767.1 | - | 12 | 1128 | |||
AK091317.1 | - | 14 | 2770 | |||
AK296546.1 | - | 6 | 788 | |||
AK315099.1 | - | 11 | 1202 | |||
AL832778.1 | - | 13 | 2637 | |||
BC075044.2 | - | 11 | 1152 | 30915575 | ||
BC075045.2 | - | 12 | 1155 | 30915359 | ||
BX647406.1 | - | 6 | 821 | |||
GU727648.1 | - | 14 | 2670 |