Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015332.3 | NP_056147.2 | 5 | 1114 | ||
XM_011515247.2 | XP_011513549.1 | 5 | 1203 | |||
XM_017011909.1 | XP_016867398.1 | 6 | 1140 | |||
XM_017011910.1 | XP_016867399.1 | 6 | 998 | |||
GenBank mRNA | AB028991.1 | - | 5 | 908 | ||
AK024226.1 | - | 4 | 622 | |||
AK127163.1 | - | 515 | ||||
AK307613.1 | - | 3 | 421 | |||
BC003691.2 | - | 5 | 975 | 3453305 | ||
BC011673.2 | - | 4 | 734 | |||
BC017657.1 | - | 6 | 1063 | |||
BC035014.2 | - | 5 | 1012 | 4812229 | ||
EU350932.1 | - | 5 | 932 |