Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015332.3 | NP_056147.2 | 6 | 1806 | ||
XM_017011909.1 | XP_016867398.1 | 7 | 1832 | |||
XM_017011910.1 | XP_016867399.1 | 7 | 1690 | |||
GenBank mRNA | AB028991.1 | - | 6 | 1600 | ||
AK024226.1 | - | 5 | 1313 | |||
AK025555.1 | - | 1 | 196 | |||
AK026126.1 | - | 1 | 601 | |||
AK124729.1 | - | 1 | 1709 | |||
AK127163.1 | - | 1207 | ||||
AK307613.1 | - | 4 | 1111 | |||
BC003691.2 | - | 6 | 1667 | 3453305 | ||
BC011673.2 | - | 5 | 1426 | |||
BC017657.1 | - | 7 | 1755 | |||
BC035014.2 | - | 6 | 1704 | 4812229 |