Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015332.3 | NP_056147.2 | 6 | 3035 | ||
XM_017011909.1 | XP_016867398.1 | 7 | 3061 | |||
XM_017011910.1 | XP_016867399.1 | 7 | 2919 | |||
GenBank mRNA | AB028991.1 | - | 6 | 2829 | ||
AK024226.1 | - | 5 | 2542 | |||
AK025555.1 | - | 1 | 1425 | |||
AK026126.1 | - | 1 | 1830 | |||
AK124729.1 | - | 1 | 2938 | |||
AK127163.1 | - | 2436 | ||||
BC003691.2 | - | 6 | 2896 | 3453305 | ||
BC011673.2 | - | 5 | 2655 | |||
BC017657.1 | - | 7 | 2984 | |||
BC035014.2 | - | 6 | 2933 | 4812229 |