Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015395.2 | NP_056210.1 | 9 | 1325 | ||
XM_005250253.3 | XP_005250310.1 | 9 | 1297 | |||
XM_017011938.1 | XP_016867427.1 | 9 | 1309 | |||
GenBank mRNA | AB037779.1 | - | 7 | 901 | ||
AK057048.1 | - | 8 | 1052 | |||
AK292996.1 | - | 9 | 1227 | |||
BC053591.1 | - | 9 | 1230 | 5502429 |