Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015419.3 | NP_056234.2 | 5 | 1467 | ||
XM_005274485.2 | XP_005274542.1 | 5 | 1922 | |||
GenBank mRNA | AF245505.1 | - | 4 | 1310 | ||
BC011846.2 | - | 5 | 1464 | |||
BC064986.1 | - | 5 | 1464 | |||
BC080586.1 | - | 5 | 1464 |