Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042594.1 | NP_001036059.1 | 7 | 881 | ||
NM_015450.2 | NP_056265.2 | 8 | 1012 | |||
NR_003102.1 | 8 | 1012 | ||||
NR_003103.1 | 8 | 1012 | ||||
NR_003104.1 | 8 | 1012 | ||||
XM_006715917.3 | XP_006715980.1 | 8 | 1002 | |||
XM_011516006.1 | XP_011514308.1 | 9 | 1124 | |||
XM_011516007.1 | XP_011514309.1 | 9 | 1123 | |||
XM_017011941.1 | XP_016867430.1 | 8 | 1010 | |||
XM_017011942.1 | XP_016867431.1 | 7 | 872 | |||
GenBank mRNA | AK001230.1 | - | 4 | 434 | ||
AK001935.1 | - | 5 | 541 | |||
AK022580.1 | - | 8 | 849 | |||
AK302263.1 | - | 8 | 1167 | |||
AY823520.1 | - | 4 | 422 | |||
AY823521.1 | - | 4 | 422 | |||
AY823522.1 | - | 4 | 422 | |||
AY823523.1 | - | 3 | 291 | |||
AY823524.1 | - | 4 | 422 | |||
BC002923.2 | - | 5 | 468 | 3955573 |