Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015621.2 | NP_056436.2 | 5, 6 | 524 | ||
GenBank mRNA | AK023767.1 | - | 4, 5 | 302 | ||
AK225826.1 | - | 5, 6 | 494 | |||
AK292841.1 | - | 5, 6 | 524 | |||
AL713742.1 | - | 4, 5 | 448 | |||
BC016647.1 | - | 4, 5 | 362 | |||
BM927471.1 | - | 41 |